siRNA Details
Gene Symbol: Clcn7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011930.3 | NP_036060.1 | 21 | 2045 | ||
GenBank mRNA | AK030444.1 | - | 21 | 2046 | ||
AK032674.1 | - | 8 | 775 | |||
AK128981.1 | - | 20 | 1971 | |||
AK150395.1 | - | 21 | 1956 | |||
AK188196.1 | - | 2 | 113 | |||
BC050907.1 | - | 21 | 2017 | 5698121 | ||
BC053049.1 | - | 21 | 2017 | 5698121 | ||
BC054799.1 | - | 21 | 2025 | 6849205 |
