siRNA Details
Gene Symbol: Fgd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159538.1 | NP_001153010.1 | 14 | 1727 | ||
NM_013710.4 | NP_038738.2 | 14 | 1724 | |||
GenBank mRNA | AF017368.1 | - | 14 | 1666 | ||
AK042260.1 | - | 14 | 1725 | |||
AK156151.1 | - | 14 | 1724 | |||
BC021845.1 | - | 11 | 1179 | 4016697 | ||
DQ344523.1 | - | 14 | 1548 |
