siRNA Details
Gene Symbol: Fgd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159538.1 | NP_001153010.1 | 8 | 1195 | ||
NM_013710.4 | NP_038738.2 | 8 | 1192 | |||
GenBank mRNA | AF017368.1 | - | 8 | 1134 | ||
AK042260.1 | - | 8 | 1193 | |||
AK156151.1 | - | 8 | 1192 | |||
BC021845.1 | - | 5 | 647 | 4016697 | ||
DQ344523.1 | - | 8 | 1016 |
