siRNA Details
Gene Symbol: Map2k4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009157.4 | NP_033183.1 | 9 | 959 | ||
GenBank mRNA | AK036928.1 | - | 9 | 963 | ||
AK044534.1 | - | 9 | 951 | |||
AK044698.1 | - | 9 | 951 | |||
AK139777.1 | - | 10 | 982 | |||
BC029833.1 | - | 7 | 710 | 3962163 | ||
U18310.1 | - | 9 | 934 |
