siRNA Details
Gene Symbol: Map3k1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011945.2 | NP_036075.2 | 19 | 4331 | ||
GenBank mRNA | AF117340.1 | - | 19 | 4331 | ||
L13103.1 | - | 9 | 2340 | |||
U23470.1 | - | 6 | 1932 |
