siRNA Details
Gene Symbol: Map3k7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009316.1 | NP_033342.1 | 5 | 548 | ||
NM_172688.3 | NP_766276.1 | 5 | 548 | |||
GenBank mRNA | AK046565.1 | - | 5 | 514 | ||
AK053911.1 | - | 5 | 534 | |||
AK137046.1 | - | 5 | 549 | |||
AK145302.1 | - | 5 | 559 | |||
AK159277.1 | - | 5 | 512 | |||
AK211827.1 | - | 3 | 146 | |||
BC006665.1 | - | 5 | 539 | 3499247 | ||
D76446.1 | - | 5 | 548 |
