siRNA Details
Gene Symbol: Map3k8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007746.2 | NP_031772.1 | 4 | 871 | ||
GenBank mRNA | AK140334.1 | - | 4 | 872 | ||
AK149624.1 | - | 4 | 872 | |||
AK149846.1 | - | 4 | 872 | |||
AK157123.1 | - | 4 | 838 | |||
BC125286.1 | - | 3 | 772 | 40129801 | ||
BC137922.1 | - | 3 | 772 | 8860941 | ||
D13759.1 | - | 4 | 871 |
