siRNA Details
Gene Symbol: Slc27a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011978.2 | NP_036108.2 | 10 | 1950 | ||
GenBank mRNA | AF033031.1 | - | 10 | 1831 | ||
AF072757.1 | - | 10 | 1828 | |||
AI326155.1 | - | 1 | 356 | |||
AJ223958.1 | - | 10 | 1868 | |||
AK144070.1 | - | 10 | 1864 | |||
BC013442.1 | - | 10 | 1933 | 4236616 | ||
BC022170.1 | - | 10 | 1929 | 4159081 | ||
BC024735.1 | - | 10 | 1919 | 5123650 |
