siRNA Details
Gene Symbol: Lgsn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153601.1 | NP_705829.1 | 5 | 597 | ||
GenBank mRNA | AF545851.1 | - | 5 | 597 | ||
BC117745.1 | - | 5 | 597 | 40090427 | ||
BC117746.1 | - | 5 | 597 | 40090429 | ||
BC128067.1 | - | 5 | 597 | 40090416 |
