siRNA Details
Gene Symbol: Sh3pxd2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177364.3 | NP_796338.2 | 1, 2 | 149 | ||
GenBank mRNA | AB430861.1 | - | 1, 2 | 71 | ||
AK042261.1 | - | 1, 2 | 135 | |||
AK043324.1 | - | 1, 2 | 121 | |||
AK089330.1 | - | 1, 2 | 121 | |||
AK090119.1 | - | 1, 2 | 142 | |||
AK154898.1 | - | 1, 2 | 107 | |||
AK163471.1 | - | 1, 2 | 121 | |||
AK169978.1 | - | 1, 2 | 141 | |||
AK171384.1 | - | 1, 2 | 121 | |||
AK220175.1 | - | 1, 2 | 138 | |||
BC115711.1 | - | 1, 2 | 118 | |||
BC115764.1 | - | 1, 2 | 118 | |||
BC141306.1 | - | 1, 2 | 121 | 9055918 | ||
BY197208.1 | - | 1, 2 | 149 |
