siRNA Details
Gene Symbol: Abcd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011994.2 | NP_036124.2 | 1 | 653 | ||
GenBank mRNA | AK051445.1 | - | 1 | 627 | ||
AK082588.1 | - | 1 | 625 | |||
AK087653.1 | - | 1 | 272 | |||
AK161027.1 | - | 1 | 314 | |||
BC019187.1 | - | 1 | 597 | 5027149 | ||
Z48670.1 | - | 1 | 657 |
