siRNA Details
Gene Symbol: Wdfy2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175546.4 | NP_780755.2 | 2 | 421 | ||
GenBank mRNA | AK045072.1 | - | 2 | 226 | ||
AK086275.1 | - | 2 | 434 | |||
AK131958.1 | - | 2 | 420 | |||
AK151817.1 | - | 2 | 377 | |||
AK169499.1 | - | 2 | 398 | |||
BC116632.1 | - | 2 | 367 | 40056912 |
