siRNA Details
Gene Symbol: Cops7a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012003.2 | NP_036133.1 | 8 | 966 | ||
GenBank mRNA | AF071316.1 | - | 8 | 968 | ||
BC003724.1 | - | 7 | 847 | 3593979 | ||
CF898433.1 | - | 4 | 462 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001164091, AI603423.
Gene Ontology Categories:
