siRNA Details
Gene Symbol: Trpc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF139923.1 | - | 3 | 1016 | ||
AK135153.1 | - | 3 | 1205 | |||
AK137418.1 | - | 3 | 1217 | |||
BC117995.1 | - | 3 | 1016 | 40096022 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Trpc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF139923.1 | - | 3 | 1016 | ||
AK135153.1 | - | 3 | 1205 | |||
AK137418.1 | - | 3 | 1217 | |||
BC117995.1 | - | 3 | 1016 | 40096022 |