siRNA Details
Gene Symbol: Zscan20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177758.4 | NP_808426.2 | 8 | 2727 | ||
GenBank mRNA | AK047820.1 | - | 8 | 2367 | ||
AK144249.1 | - | 8 | 2728 | |||
AK153688.1 | - | 8 | 2664 | |||
AK153845.1 | - | 8 | 2664 | |||
BC023282.1 | - | 1 | 669 | |||
BC065079.1 | - | 4 | 1317 | 6825639 | ||
BC076602.1 | - | 8 | 2643 | 30611085 |
