siRNA Details
Gene Symbol: Zscan20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177758.4 | NP_808426.2 | 7 | 1811 | ||
GenBank mRNA | AK047820.1 | - | 7 | 1451 | ||
AK144249.1 | - | 7 | 1812 | |||
AK153688.1 | - | 7 | 1748 | |||
AK153845.1 | - | 7 | 1748 | |||
BC065079.1 | - | 3 | 401 | 6825639 | ||
BC076602.1 | - | 7 | 1727 | 30611085 |
