siRNA Details
Gene Symbol: Lhfpl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081231.2 | NP_001074700.2 | 2 | 597 | ||
NM_029990.1 | NP_084266.1 | 2 | 597 | |||
GenBank mRNA | AK033906.1 | - | 2 | 571 | ||
AK076730.1 | - | 2 | 262 |
