siRNA Details
Gene Symbol: A930003O13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK044256.1 | - | 1 | 354 | ||
BC030424.1 | - | 1 | 340 |
Gene Symbol: Lhfpl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081231.2 | NP_001074700.2 | 1 | 359 | ||
NM_029990.1 | NP_084266.1 | 1 | 359 | |||
GenBank mRNA | AK020916.1 | - | 1 | 328 | ||
AK033906.1 | - | 1 | 333 |

More Information
Other Species Targeted

Gene Ontology Categories:
