siRNA Details
Gene Symbol: Polk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012048.2 | NP_036178.1 | 12, 13 | 1712 | ||
GenBank mRNA | AB027563.1 | - | 12, 13 | 1700 | ||
AF163571.1 | - | 12, 13 | 1712 | |||
AY769920.1 | - | 11, 12 | 1349 | |||
AY769921.1 | - | 10, 11 | 1286 | |||
AY769923.1 | - | 8, 9 | 1086 | |||
AY769924.1 | - | 8, 9 | 994 | |||
BC052820.1 | - | 9, 10 | 1377 | 30057338 |
