siRNA Details
Gene Symbol: G3bp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013716.2 | NP_038744.1 | 12 | 1416 | ||
GenBank mRNA | AB001927.1 | - | 12 | 1416 | ||
AK141689.1 | - | 11 | 1373 | |||
AK200144.1 | - | 1 | 14 | |||
AK220230.1 | - | 12 | 1470 | |||
BC021156.1 | - | 12 | 1428 | 4020362 |
