siRNA Details
Gene Symbol: G3bp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013716.2 | NP_038744.1 | 5 | 477 | ||
GenBank mRNA | AB001927.1 | - | 5 | 477 | ||
AK141689.1 | - | 5 | 544 | |||
AK208718.1 | - | 2 | 94 | |||
AK220230.1 | - | 5 | 531 | |||
BC021156.1 | - | 5 | 489 | 4020362 |
