siRNA Details
Gene Symbol: Mthfd1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170785.1 | NP_001164256.1 | 12 | 1509 | ||
NM_001170786.1 | NP_001164257.1 | 12 | 1509 | |||
NM_172308.4 | NP_758512.3 | 12 | 1509 | |||
GenBank mRNA | AK028211.1 | - | 12 | 1460 | ||
AK036284.1 | - | 12 | 1455 | |||
AK038579.1 | - | 12 | 1467 | |||
AK075931.1 | - | 12 | 1435 | |||
AK160025.1 | - | 12 | 1441 | |||
BC030437.1 | - | 8 | 843 | 5374413 | ||
BC049936.1 | - | 8 | 832 | |||
BC058265.1 | - | 12 | 1351 |
