siRNA Details
Gene Symbol: Agbl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178755.3 | NP_848870.2 | 9 | 1346 | ||
GenBank mRNA | AK029744.1 | - | 9 | 1379 | ||
AK029932.1 | - | 9 | 1347 | |||
AK030138.1 | - | 9 | 1334 | |||
AK030182.1 | - | 9 | 1393 | |||
AK040259.1 | - | 9 | 1282 | |||
AK079691.1 | - | 9 | 1283 | |||
DQ867026.1 | - | 9 | 1379 | |||
DQ867027.1 | - | 9 | 1379 | |||
DQ867028.1 | - | 9 | 1379 | |||
DQ867029.1 | - | 9 | 1379 | |||
DQ867030.1 | - | 8 | 1325 |
