siRNA Details
Gene Symbol: Cars | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252593.1 | NP_001239522.1 | 11, 12 | 1229 | ||
NM_013742.5 | NP_038770.3 | 12, 13 | 1478 | |||
GenBank mRNA | AB015589.1 | - | 9, 10 | 1012 | ||
AJ276796.1 | - | 12, 13 | 1445 | |||
AK033328.1 | - | 12, 13 | 1445 | |||
AK037227.1 | - | 11, 12 | 1197 | |||
AK047027.1 | - | 11, 12 | 1197 | |||
AK077558.1 | - | 11, 12 | 1195 | |||
AK133205.1 | - | 11, 12 | 1196 | |||
AK135433.1 | - | 12, 13 | 1448 | |||
AK152866.1 | - | 11, 12 | 1202 | |||
AK198552.1 | - | 1, 2 | 38 | |||
BC030473.1 | - | 8, 9 | 914 | |||
BC054717.1 | - | 12, 13 | 1427 | 5686364 | ||
BC058954.1 | - | 12, 13 | 1430 | 5707198 |
