siRNA Details
Gene Symbol: Cars | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252593.1 | NP_001239522.1 | 3, 4 | 296 | ||
NM_013742.5 | NP_038770.3 | 4, 5 | 545 | |||
GenBank mRNA | AB015589.1 | - | 1, 2 | 79 | ||
AJ276796.1 | - | 4, 5 | 512 | |||
AK033328.1 | - | 4, 5 | 512 | |||
AK037227.1 | - | 3, 4 | 264 | |||
AK047027.1 | - | 3, 4 | 263 | |||
AK077558.1 | - | 3, 4 | 262 | |||
AK133205.1 | - | 3, 4 | 263 | |||
AK135433.1 | - | 4, 5 | 515 | |||
AK152866.1 | - | 3, 4 | 269 | |||
BC054717.1 | - | 4, 5 | 494 | 5686364 | ||
BC058954.1 | - | 4, 5 | 497 | 5707198 |
