siRNA Details
Gene Symbol: Adamts13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001322.2 | NP_001001322.1 | 1 | 189 | ||
NM_001290463.1 | NP_001277392.1 | 1 | 189 | |||
NM_001290464.1 | NP_001277393.1 | 1 | 189 | |||
NM_001290465.1 | NP_001277394.1 | 1 | 189 | |||
GenBank mRNA | AB071302.2 | - | 1 | 189 | ||
AB112362.1 | - | 1 | 68 | |||
AK157459.1 | - | 1 | 113 | |||
EU034706.1 | - | 1 | 68 |
