siRNA Details
Gene Symbol: Zcchc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138590.4 | NP_613056.2 | 7 | 1177 | ||
GenBank mRNA | AK013175.1 | - | 1 | 661 | ||
AK050430.1 | - | 1 | 1869 | |||
BC050200.1 | - | 6 | 1052 | |||
BC054405.1 | - | 1 | 380 |
