siRNA Details
Gene Symbol: Zcchc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138590.4 | NP_613056.2 | 9 | 1604 | ||
GenBank mRNA | AK013088.1 | - | 1 | 133 | ||
AK013175.1 | - | 3 | 1088 | |||
AK050430.1 | - | 3 | 2296 | |||
BC050200.1 | - | 8 | 1479 | |||
BC054405.1 | - | 3 | 807 |
