siRNA Details
Gene Symbol: Gfm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138591.2 | NP_613057.2 | 7 | 1001 | ||
GenBank mRNA | AF315511.1 | - | 7 | 968 | ||
AK033214.1 | - | 7 | 1000 | |||
BC013093.1 | - | 7 | 1004 | 3584730 | ||
BC031772.1 | - | 7 | 968 | 4239980 |
