siRNA Details
Gene Symbol: Usp39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138592.4 | NP_613058.1 | 12 | 1598 | ||
GenBank mRNA | AK010443.1 | - | 12 | 1608 | ||
AK018595.1 | - | 12 | 1599 | |||
AK033105.1 | - | 12 | 1595 | |||
AK045062.1 | - | 12 | 1595 | |||
AK088354.1 | - | 12 | 1610 | |||
AK165625.1 | - | 12 | 1579 | |||
AK167668.1 | - | 12 | 1586 | |||
AK191104.1 | - | 1 | 25 | |||
AK211034.1 | - | 1 | 25 | |||
AK216737.1 | - | 1 | 25 | |||
BC026983.1 | - | 12 | 1573 | 5365862 |
