siRNA Details
Gene Symbol: Limd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013860.2 | NP_038888.2 | 1 | 1541 | ||
GenBank mRNA | AJ132409.1 | - | 1 | 1530 | ||
AK004806.1 | - | 1 | 845 | |||
AK047198.1 | - | 1 | 1535 | |||
AK083970.1 | - | 1 | 1502 | |||
BC056449.1 | - | 1 | 1416 | 5703026 |
