siRNA Details
Gene Symbol: Tpk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013861.3 | NP_038889.1 | 7 | 513 | ||
GenBank mRNA | AB027568.1 | - | 7 | 514 | ||
AK028431.1 | - | 7 | 536 | |||
AK033192.1 | - | 5 | 617 | |||
AK034521.1 | - | 523 | ||||
AK035044.1 | - | 7 | 514 | |||
AK052225.1 | - | 7 | 505 | |||
AK087844.1 | - | 7 | 617 | |||
BC015246.1 | - | 7 | 530 | 4221866 |
