siRNA Details
Gene Symbol: Tpk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013861.3 | NP_038889.1 | 8, 9 | 674 | ||
GenBank mRNA | AB027568.1 | - | 8, 9 | 675 | ||
AK028431.1 | - | 8, 9 | 697 | |||
AK033192.1 | - | 6, 7 | 778 | |||
AK034521.1 | - | 684 | ||||
AK035044.1 | - | 8, 9 | 675 | |||
AK052225.1 | - | 8, 9 | 666 | |||
AK087844.1 | - | 8, 9 | 779 | |||
AK136486.1 | - | 7, 8 | 528 | |||
BC015246.1 | - | 8, 9 | 691 | 4221866 | ||
BC023354.1 | - | 7, 8 | 503 | 5253545 |
