siRNA Details
Gene Symbol: Zbtb18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012330.1 | NP_001012330.1 | 2 | 404 | ||
NM_013915.3 | NP_038943.3 | 1 | 1510 | |||
GenBank mRNA | AF140224.1 | - | 1 | 1512 | ||
AK147200.1 | - | 2 | 361 | |||
BC050909.1 | - | 2 | 431 | |||
BC054529.1 | - | 1 | 512 | 5721393 | ||
BC054742.1 | - | 1 | 512 | 5721393 | ||
BC089333.1 | - | 2 | 412 | |||
BU701724.1 | - | 1 | 512 | |||
CF539422.1 | - | 2 | 412 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_022678, XM_006250331, XM_006250328, XM_008769799, XM_008769796, and others.
Gene Ontology Categories:
