siRNA Details
Gene Symbol: Usp25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013918.2 | NP_038946.2 | 18 | 2326 | ||
GenBank mRNA | AF170563.1 | - | 18 | 2472 | ||
AK077769.1 | - | 3 | 499 | |||
BC048171.1 | - | 18 | 2326 | 6822095 | ||
BC063059.1 | - | 18 | 2326 | 6822095 |
