siRNA Details
Gene Symbol: Mapk8ip3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163447.1 | NP_001156919.1 | 26 | 3277 | ||
NM_001163448.1 | NP_001156920.1 | 25 | 3235 | |||
NM_001163449.1 | NP_001156921.1 | 27 | 3211 | |||
NM_001163450.1 | NP_001156922.1 | 26 | 3187 | |||
NM_001163451.1 | NP_001156923.1 | 26 | 3184 | |||
NM_001163453.1 | NP_001156925.1 | 25 | 3169 | |||
NM_013931.4 | NP_038959.2 | 26 | 3280 | |||
GenBank mRNA | AB005662.1 | - | 26 | 3167 | ||
AB043123.1 | - | 27 | 3088 | |||
AB043124.1 | - | 26 | 3157 | |||
AB043125.1 | - | 26 | 3154 | |||
AB093282.2 | - | 26 | 3139 | |||
AF178636.1 | - | 25 | 3152 | |||
AF178637.1 | - | 26 | 3273 | |||
AK039130.1 | - | 8 | 907 | |||
AK134797.1 | - | 5 | 355 | |||
BC060603.1 | - | 27 | 3146 | 5706805 | ||
BC065105.1 | - | 25 | 3159 | 30356817 | ||
BC068312.1 | - | 26 | 3258 | 30100914 |
