siRNA Details
Gene Symbol: Nol4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161483.1 | NP_001154955.1 | 9 | 1332 | ||
NM_199024.2 | NP_950189.2 | 8 | 2910 | |||
GenBank mRNA | AK029800.1 | - | 4 | 644 | ||
AK031008.1 | - | 8 | 2910 | |||
AK031753.1 | - | 2 | 325 | |||
AK086574.1 | - | 5 | 836 | |||
BC056377.1 | - | 9 | 1325 | 6850243 |

More Information
Other Species Targeted

Gene Ontology Categories:
