siRNA Details
Gene Symbol: Nol4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161483.1 | NP_001154955.1 | 4, 5 | 482 | ||
NM_199024.2 | NP_950189.2 | 4, 5 | 2252 | |||
GenBank mRNA | AK031008.1 | - | 4, 5 | 2252 | ||
BC056377.1 | - | 4, 5 | 475 | 6850243 |
