siRNA Details
Gene Symbol: Atg2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029654.4 | NP_083930.5 | 8 | 1553 | ||
GenBank mRNA | AK032840.1 | - | 6 | 819 | ||
AK083213.1 | - | 8 | 1513 | |||
AK085790.1 | - | 8 | 1499 | |||
AK086802.1 | - | 8 | 1554 | |||
BC062182.1 | - | 8 | 1523 | 6505402 |
