siRNA Details
Gene Symbol: Zfp750 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178763.4 | NP_848878.1 | 2 | 505 | ||
GenBank mRNA | AK030690.1 | - | 2 | 460 | ||
AK037185.1 | - | 2 | 466 | |||
AK037240.1 | - | 2 | 451 | |||
AK076268.1 | - | 2 | 497 | |||
AK140792.1 | - | 497 | ||||
BC150905.1 | - | 1 | 359 | 9087816 |
