siRNA Details
Gene Symbol: Hif1an | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176958.3 | NP_795932.2 | 3 | 497 | ||
GenBank mRNA | AK043636.1 | - | 3 | 483 | ||
AK141867.1 | - | 3 | 483 | |||
AK154774.1 | - | 3 | 497 | |||
BC130013.1 | - | 3 | 501 | 40140349 | ||
BY713373.1 | - | 3 | 483 | |||
CN535711.1 | - | 2 | 81 |
