siRNA Details
Gene Symbol: Hif1an | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176958.3 | NP_795932.2 | 2 | 312 | ||
GenBank mRNA | AK043636.1 | - | 2 | 298 | ||
AK084960.1 | - | 2 | 307 | |||
AK141867.1 | - | 2 | 298 | |||
AK154774.1 | - | 2 | 312 | |||
BC130013.1 | - | 2 | 316 | 40140349 | ||
BY122345.1 | - | 2 | 316 | |||
BY713373.1 | - | 2 | 298 |

More Information
Other Species Targeted

Gene Ontology Categories:
