siRNA Details
Gene Symbol: Galm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176963.4 | NP_795937.1 | 5 | 965 | ||
GenBank mRNA | AK040998.1 | - | 5 | 966 | ||
AK158120.1 | - | 5 | 764 | |||
AK161687.1 | - | 5 | 764 | |||
AK170014.1 | - | 5 | 767 | |||
BC028818.1 | - | 5 | 768 | 5135570 |
