siRNA Details
Gene Symbol: Agmo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178767.5 | NP_848882.2 | 5 | 956 | ||
GenBank mRNA | AK039191.1 | - | 5 | 527 | ||
AK040702.1 | - | 5 | 957 | |||
AK050246.1 | - | 5 | 817 | |||
AK163155.1 | - | 5 | 737 |

More Information
Other Species Targeted

Gene Ontology Categories:
