siRNA Details
Gene Symbol: Ankar | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176980.4 | NP_795954.2 | 10 | 2149 | ||
GenBank mRNA | AK030065.1 | - | 10 | 2150 | ||
AK077036.1 | - | 9 | 1538 | |||
BC132415.1 | - | 10 | 2124 | 40130692 | ||
BC145253.1 | - | 10 | 2121 | 9053805 |
