siRNA Details
Gene Symbol: Ankar | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176980.4 | NP_795954.2 | 18, 19 | 3620 | ||
GenBank mRNA | AK030065.1 | - | 18, 19 | 3621 | ||
AK077036.1 | - | 17, 18 | 3010 | |||
BC132415.1 | - | 18, 19 | 3595 | 40130692 | ||
BC145253.1 | - | 18, 19 | 3592 | 9053805 |
