siRNA Details
Gene Symbol: Ankar | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176980.4 | NP_795954.2 | 19 | 3728 | ||
GenBank mRNA | AK030065.1 | - | 19 | 3729 | ||
AK077036.1 | - | 18 | 3118 | |||
BC132415.1 | - | 19 | 3703 | 40130692 | ||
BC145253.1 | - | 19 | 3700 | 9053805 |
