siRNA Details
Gene Symbol: Igf2bp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183029.2 | NP_898850.2 | 12 | 1669 | ||
GenBank mRNA | AK044984.1 | - | 11 | 1185 | ||
AK049196.1 | - | 12 | 1365 | |||
AK077404.1 | - | 12 | 1350 | |||
AK159654.1 | - | 12 | 1402 | |||
AK170531.1 | - | 11 | 1173 | |||
AY531659.1 | - | 12 | 1333 | |||
BC023758.1 | - | 12 | 1367 | |||
BC054552.1 | - | 12 | 1389 | |||
BU708344.1 | - | 2 | 114 |
