siRNA Details
Gene Symbol: Auts2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177047.3 | NP_796021.2 | 2 | 905 | ||
GenBank mRNA | AK052418.1 | - | 2 | 967 | ||
AK077988.1 | - | 2 | 1362 | |||
AK147478.1 | - | 1904 | ||||
BC096021.1 | - | 2 | 904 | |||
BC151031.1 | - | 2 | 919 | |||
BC151047.1 | - | 2 | 918 |
