siRNA Details
Gene Symbol: Auts2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177047.3 | NP_796021.2 | 2, 3 | 1045 | ||
GenBank mRNA | AK052418.1 | - | 2, 3 | 1107 | ||
AK085946.1 | - | 2, 3 | 736 | |||
AK147478.1 | - | 2044 | ||||
BC096021.1 | - | 2, 3 | 1044 | |||
BC151031.1 | - | 2, 3 | 1059 | |||
BC151047.1 | - | 2, 3 | 1058 |
